Ethyl 4-methoxy-α-oxo-1-naphthaleneacetate ≥95%

Supplier: AOBChem USA

Warning

368679-5G 368679-25G 368679-1G 368679-500MG
77628-856EA 1484.23 USD
77628-856 77635-710 77638-386 77646-412
Ethyl 4-methoxy-α-oxo-1-naphthaleneacetate ≥95%
Ethyl 4-methoxy-α-oxo-1-naphthaleneacetate

Precaución: For Research Use Only

Formula: 258.27 g/mol
MW: C₁₅H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD16284934
CAS Number: 22531-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR