Cyclohexyl(4-fluoro-3,5-dimethylphenyl)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
367762-5G
367762-25G
367762-1G
367762-500MG
77628-852EA
1484.23
USD
77628-852
77635-708
77638-384
77650-412
Cyclohexyl(4-fluoro-3,5-dimethylphenyl)methanone ≥97%
Cyclohexyl(4-fluoro-3,5-dimethylphenyl)methanone
Precaución: For Research Use Only
Formula:
234.309 g/mol MW: C₁₅H₁₉FO Storage Temperature: Ambient |
CAS Number:
1490535-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...