(4-Chloro-3,5-dimethylphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

367757-5G 367757-25G 367757-1G 367757-500MG
77632-964EA 1441 USD
77632-964 77655-438 77642-390 77646-406
(4-Chloro-3,5-dimethylphenyl)(cyclopropyl)methanol ≥95%
(4-Chloro-3,5-dimethylphenyl)(cyclopropyl)methanol

Precaución: For Research Use Only

Formula: 210.7 g/mol
MW: C₁₂H₁₅ClO
Storage Temperature: Ambient
CAS Number: 3037693-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR