1-((3-Bromobenzyl)oxy)-2-(tert-butyl)-4-ethylbenzene ≥97%

Supplier: AOBChem USA

Warning

367025-5G 367025-1G 367025-25G 367025-500MG
77628-848EA 1484.23 USD
77628-848 77638-380 77658-966 77650-408
1-((3-Bromobenzyl)oxy)-2-(tert-butyl)-4-ethylbenzene ≥97%
1-((3-Bromobenzyl)oxy)-2-(tert-butyl)-4-ethylbenzene

Precaución: For Research Use Only

Formula: 347.2893 g/mol
MW: C₁₉H₂₃BrO
Storage Temperature: Ambient
MDL Number: MFCD35194330
CAS Number: 1481359-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR