1-((4-Bromobenzyl)oxy)-2-(tert-butyl)-4-ethylbenzene ≥97%

Supplier: AOBChem USA

Warning

367024-5G 367024-1G 367024-500MG
77655-432EA 1998 USD
77655-432 77642-384 77650-406
1-((4-Bromobenzyl)oxy)-2-(tert-butyl)-4-ethylbenzene ≥97%
1-((4-Bromobenzyl)oxy)-2-(tert-butyl)-4-ethylbenzene

Precaución: For Research Use Only

Formula: 347.2893 g/mol
MW: C₁₉H₂₃BrO
Storage Temperature: Ambient
MDL Number: MFCD35194329
CAS Number: 1455651-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR