2-(Benzyloxy)-1-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

36601-5G 36601-25G 36601-1G 36601-500MG
77634-934EA 523 USD
77634-934 77655-426 77642-382 77646-396
2-(Benzyloxy)-1-naphthoic acid ≥97%
2-(Benzyloxy)-1-naphthoic acid

Precaución: For Research Use Only

Formula: 278.302 g/mol
MW: C₁₈H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD04578310
CAS Number: 93655-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR