1-(3-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

365397-5G 365397-25G 365397-1G 365397-500MG
77628-842EA 1484.23 USD
77628-842 77655-422 77646-388 77646-390
1-(3-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)ethanol ≥95%
1-(3-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)ethanol

Precaución: For Research Use Only

Formula: C₉H₇BrF₄O
MW: 287.049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD35193675
CAS Number: 3002429-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR