1-(3-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)propan-1-one ≥95%
Supplier: AOBChem USA
|
Warning
|
365394-5G
365394-1G
365394-25G
365394-500MG
77628-840EA
1484.23
USD
77628-840
77638-374
77655-420
77650-404
1-(3-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)propan-1-one ≥95%
1-(3-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)propan-1-one
Precaución: For Research Use Only
Formula:
C₁₀H₇BrF₄O Storage Temperature: Ambient |
MDL Number:
MFCD35193672 CAS Number: 3002533-25-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...