1-(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)-2-methylpropan-1-one ≥95%

Supplier: AOBChem USA

Warning

365343-5G 365343-1G 365343-500MG 365343-25G
77628-838EA 1667.57 USD
77628-838 77638-372 77652-710 77655-418
1-(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)-2-methylpropan-1-one ≥95%
1-(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)-2-methylpropan-1-one

Precaución: For Research Use Only

Formula: 268.635 g/mol
MW: C₁₁H₉ClF₄O
Storage Temperature: Ambient
MDL Number: MFCD35193646
CAS Number: 3002493-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR