(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)(cyclohexyl)methanone ≥95%

Supplier: AOBChem USA

Warning

365321-5G 365321-25G 365321-1G 365321-500MG
77632-960EA 1441 USD
77632-960 77658-960 77642-380 77650-402
(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)(cyclohexyl)methanone ≥95%
(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)(cyclohexyl)methanone

Precaución: For Research Use Only

Formula: C₁₄H₁₃ClF₄O
Storage Temperature: Ambient
MDL Number: MFCD35193626
CAS Number: 3002494-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR