(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
Supplier: AOBChem USA
|
Warning
|
365313-5G
365313-1G
365313-25G
365313-500MG
77632-958EA
1441
USD
77632-958
77638-370
77658-958
77650-400
(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)(methyl)sulphane
Precaución: For Research Use Only
Formula:
244.637 g/mol MW: C₈H₅ClF₄S Storage Temperature: Ambient |
MDL Number:
MFCD35193618 CAS Number: 2711574-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...