2-(6-Chloro-2-fluoro-3-(methylthio)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

365298-5G 365298-1G 365298-25G 365298-500MG
77628-836EA 1484.23 USD
77628-836 77638-368 77658-956 77650-398
2-(6-Chloro-2-fluoro-3-(methylthio)phenyl)propan-2-ol ≥95%
2-(6-Chloro-2-fluoro-3-(methylthio)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₁₀H₁₂ClFOS
MW: 234.718 g/mol
Storage Temperature: Ambient
MDL Number: MFCD35193603
CAS Number: 3002448-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR