(4'-Chloro-[1,1'-biphenyl]-4-yl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

364051-5G 364051-500MG 364051-25G 364051-1G
77632-954EA 1441 USD
77632-954 77652-708 77658-948 77642-374
(4'-Chloro-[1,1'-biphenyl]-4-yl)(cyclopropyl)methanol ≥95%
(4'-Chloro-[1,1'-biphenyl]-4-yl)(cyclopropyl)methanol

Precaución: For Research Use Only

Formula: 258.743 g/mol
MW: C₁₆H₁₅ClO
Storage Temperature: Ambient
MDL Number: MFCD35193169
CAS Number: 3002425-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR