(4'-Chloro-[1,1'-biphenyl]-4-yl)(ethyl)sulphane ≥95%

Supplier: AOBChem USA

Warning

364040-5G 364040-25G 364040-1G 364040-500MG
77632-952EA 1441 USD
77632-952 77658-946 77642-372 77646-382
(4'-Chloro-[1,1'-biphenyl]-4-yl)(ethyl)sulphane ≥95%
(4'-Chloro-[1,1'-biphenyl]-4-yl)(ethyl)sulphane

Precaución: For Research Use Only

Formula: C₁₄H₁₃ClS
Storage Temperature: Ambient
MDL Number: MFCD35193158
CAS Number: 3002425-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR