1-Bromo-2-fluoro-3-methyl-5-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

36331-5G 36331-25G 36331-1G 36331-500MG
77632-950EA 1273 USD
77632-950 77655-410 77642-370 77650-392
1-Bromo-2-fluoro-3-methyl-5-(trifluoromethyl)benzene ≥95%
1-Bromo-2-fluoro-3-methyl-5-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 257.023 g/mol
MW: C₈H₅BrF₄
Storage Temperature: Ambient
MDL Number: MFCD31568392
CAS Number: 2167794-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR