(2-(Benzyloxy)-5-fluorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

36252-5G 36252-25G 36252-1G 36252-500MG
77632-946EA 1127 USD
77632-946 77658-942 77642-368 77646-380
(2-(Benzyloxy)-5-fluorophenyl)methanol ≥95%
(2-(Benzyloxy)-5-fluorophenyl)methanol

Precaución: For Research Use Only

Formula: C₁₄H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD11639963
CAS Number: 331729-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR