(2,6-Dichloro-4-methylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

362073-5G 362073-1G 362073-25G 362073-500MG
77632-944EA 1441 USD
77632-944 77638-360 77655-406 77650-390
(2,6-Dichloro-4-methylphenyl)trimethylsilane ≥95%
(2,6-Dichloro-4-methylphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 233.21 g/mol
MW: C₁₀H₁₄Cl₂Si
Storage Temperature: Ambient
MDL Number: MFCD35192379
CAS Number: 3002458-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR