1-(2,6-Dichloro-4-methylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

362056-500MG 362056-5G 362056-1G
77652-704EA 452 USD
77652-704 77655-404 77642-366
1-(2,6-Dichloro-4-methylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(2,6-Dichloro-4-methylphenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 257.037 g/mol
MW: C₉H₅Cl₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD28142027
CAS Number: 1823577-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR