2',3'-Dichloro-6'-fluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

361079-5G 361079-25G
77634-932EA 121 USD
77634-932 77658-940
2',3'-Dichloro-6'-fluoroacetophenone ≥97%
2',3'-Dichloro-6'-fluoroacetophenone

Precaución: For Research Use Only

Formula: C₈H₅Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD06798121
CAS Number: 870704-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR