4-((2-(Tert-butyl)phenoxy)methyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

360901-5G 360901-25G 360901-1G 360901-500MG
77628-828EA 1565.6 USD
77628-828 77655-400 77642-364 77650-384
4-((2-(Tert-butyl)phenoxy)methyl)benzaldehyde ≥97%
4-((2-(Tert-butyl)phenoxy)methyl)benzaldehyde

Precaución: For Research Use Only

Formula: 268.3502 g/mol
MW: C₁₈H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD35192010
CAS Number: 3002451-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR