Cyclobutyl(3-methyl-5-(trimethylsilyl)phenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

360252-5G 360252-25G 360252-1G 360252-500MG
77628-822EA 1484.23 USD
77628-822 77635-704 77638-354 77650-378
Cyclobutyl(3-methyl-5-(trimethylsilyl)phenyl)methanone ≥97%
Cyclobutyl(3-methyl-5-(trimethylsilyl)phenyl)methanone

Precaución: For Research Use Only

Formula: C₁₅H₂₂OSi
Storage Temperature: Ambient
MDL Number: MFCD35191868
CAS Number: 3002507-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR