(3-Methyl-5-(trimethylsilyl)phenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

360240-5G 360240-25G 360240-1G 360240-500MG
77632-942EA 1441 USD
77632-942 77655-394 77642-360 77650-376
(3-Methyl-5-(trimethylsilyl)phenyl)(phenyl)methanol ≥97%
(3-Methyl-5-(trimethylsilyl)phenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: C₁₇H₂₂OSi
Storage Temperature: Ambient
MDL Number: MFCD35191856
CAS Number: 3002433-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR