1-(3-Methyl-5-(trimethylsilyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

360209-5G 360209-25G 360209-1G 360209-500MG
77628-814EA 1599.59 USD
77628-814 77655-388 77642-356 77650-372
1-(3-Methyl-5-(trimethylsilyl)phenyl)ethanone ≥95%
1-(3-Methyl-5-(trimethylsilyl)phenyl)ethanone

Precaución: For Research Use Only

Formula: 206.356 g/mol
MW: C₁₂H₁₈OSi
Storage Temperature: Ambient
MDL Number: MFCD35191825
CAS Number: 3002455-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR