1-Bromo-3-isopropoxy-5-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

36017-5G 36017-25G 36017-1G 36017-500MG
77634-930EA 598 USD
77634-930 77658-936 77646-368 77646-370
1-Bromo-3-isopropoxy-5-methoxybenzene ≥95%
1-Bromo-3-isopropoxy-5-methoxybenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD22205811
CAS Number: 1235566-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR