3-Benzyl-5-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

360146-5G 360146-1G 360146-25G 360146-500MG
77628-812EA 1599.59 USD
77628-812 77638-350 77655-386 77650-368
3-Benzyl-5-methylbenzaldehyde ≥95%
3-Benzyl-5-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₁₅H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD35191768
CAS Number: 1229244-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR