3-Benzyl-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

360141-5G 360141-1G 360141-500MG 360141-25G
77628-810EA 1599.59 USD
77628-810 77638-348 77652-702 77658-932
3-Benzyl-5-methylbenzoic acid ≥97%
3-Benzyl-5-methylbenzoic acid

Precaución: For Research Use Only

Formula: 226.27 g/mol
MW: C₁₅H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD35191763
CAS Number: 3002432-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR