α-(3,3-Dimethyl-1-butyn-1-yl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

359977-5G 359977-1G 359977-500MG
77628-808EA 1599.59 USD
77628-808 77642-352 77646-366
α-(3,3-Dimethyl-1-butyn-1-yl)benzenemethanol ≥97%
α-(3,3-Dimethyl-1-butyn-1-yl)benzenemethanol

Precaución: For Research Use Only

Formula: C₁₃H₁₆O
MW: 188.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28990294
CAS Number: 17474-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR