α-Ethyl-2,5-bis(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

359782-5G 359782-25G 359782-1G 359782-500MG
77628-806EA 1484.23 USD
77628-806 77635-702 77638-342 77650-362
α-Ethyl-2,5-bis(trifluoromethyl)benzenemethanol ≥95%
α-Ethyl-2,5-bis(trifluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: 272.19 g/mol
MW: C₁₁H₁₀F₆O
Storage Temperature: Ambient
MDL Number: MFCD00033355
CAS Number: 195371-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR