1-Bromo-2-(2-cyclopropylethynyl)benzene ≥97%

Supplier: AOBChem USA

Warning

358946-5G 358946-1G 358946-500MG
77628-802EA 1484.23 USD
77628-802 77642-350 77650-360
1-Bromo-2-(2-cyclopropylethynyl)benzene ≥97%
1-Bromo-2-(2-cyclopropylethynyl)benzene

Precaución: For Research Use Only

Formula: 221.09 g/mol
MW: C₁₁H₉Br
Storage Temperature: Ambient
MDL Number: MFCD35191421
CAS Number: 1224882-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR