1-(4-Bromo-3-methylphenyl)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

35892-5G 35892-1G 35892-25G 35892-500MG
77632-934EA 706 USD
77632-934 77638-338 77658-928 77646-362
1-(4-Bromo-3-methylphenyl)-1H-pyrazole ≥97%
1-(4-Bromo-3-methylphenyl)-1H-pyrazole

Precaución: For Research Use Only

Formula: C₁₀H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD15144545
CAS Number: 858523-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR