1-(2-Cyclopropylethynyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

358874-5G 358874-1G 358874-25G 358874-500MG
77632-932EA 1357 USD
77632-932 77638-336 77658-926 77650-358
1-(2-Cyclopropylethynyl)cyclopentanol ≥95%
1-(2-Cyclopropylethynyl)cyclopentanol

Precaución: For Research Use Only

Formula: C₁₀H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD35191411
CAS Number: 2459883-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR