3-Cyclopropyl-2-propynoic acid ≥95%

Supplier: AOBChem USA

Warning

358847-5G 358847-25G 358847-1G 358847-500MG
77632-930EA 712 USD
77632-930 77655-378 77642-348 77650-356
3-Cyclopropyl-2-propynoic acid ≥95%
3-Cyclopropyl-2-propynoic acid

Precaución: For Research Use Only

Formula: C₆H₆O₂
Storage Temperature: Ambient
MDL Number: MFCD03035228
CAS Number: 7358-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR