4-Cyclopropyl-3-butyn-2-one ≥95%

Supplier: AOBChem USA

Warning

358842-5G 358842-25G 358842-1G 358842-500MG
77632-928EA 1357 USD
77632-928 77658-924 77646-358 77650-354
4-Cyclopropyl-3-butyn-2-one ≥95%
4-Cyclopropyl-3-butyn-2-one

Precaución: For Research Use Only

Formula: C₇H₈O
MW: 108.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24697823
CAS Number: 681432-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR