2-(6-Methoxynaphthyl)trifluoromethyl ketone ≥97%

Supplier: AOBChem USA

Warning

35872-5G 35872-1G 35872-25G 35872-500MG
77634-928EA 456 USD
77634-928 77638-330 77655-376 77646-354
2-(6-Methoxynaphthyl)trifluoromethyl ketone ≥97%
2-(6-Methoxynaphthyl)trifluoromethyl ketone

Precaución: For Research Use Only

Formula: C₁₃H₉F₃O₂
MW: 254.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01320012
CAS Number: 886370-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR