2-(Allyloxy)-5-chlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

358286-5G 358286-25G
77634-924EA 142 USD
77634-924 77655-370
2-(Allyloxy)-5-chlorobenzaldehyde ≥97%
2-(Allyloxy)-5-chlorobenzaldehyde

Precaución: For Research Use Only

Formula: C₁₀H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD03946381
CAS Number: 152842-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR