4-Fluoro-1,1':3',1''-terphenyl ≥95%

Supplier: AOBChem USA

Warning

358063-1G 358063-5G 358063-500MG
77634-920EA 803 USD
77634-920 77655-366 77646-352
4-Fluoro-1,1':3',1''-terphenyl ≥95%
4-Fluoro-1,1':3',1''-terphenyl

Precaución: For Research Use Only

Formula: C₁₈H₁₃F
Storage Temperature: Ambient
MDL Number: MFCD35191111
CAS Number: 952483-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR