1-(4-Chloro-2-ethoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

357952-5G 357952-25G 357952-1G 357952-500MG
77632-920EA 760 USD
77632-920 77658-918 77642-340 77650-348
1-(4-Chloro-2-ethoxyphenyl)ethanone ≥95%
1-(4-Chloro-2-ethoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: C₁₀H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD15528680
CAS Number: 151945-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR