1-(2-Bromoethoxy)-3-chlorobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Benzene, 1-(2-bromoethoxy)-3-chloro-, 3-Chloro-β-bromophenetol, 3-Chloro-ω-bromophenetol

357896-5G 357896-1G 357896-25G
77634-918EA 198 USD
77634-918 77638-326 77658-916
1-(2-Bromoethoxy)-3-chlorobenzene ≥97%
1-(2-Bromoethoxy)-3-chlorobenzene

Precaución: For Research Use Only

Formula: C₈H₈BrClO
MW: 235.51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02030630
CAS Number: 6487-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR