6-Bromo-5-fluoro-3-iodo-1-methyl-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

357866-5G 357866-1G 357866-500MG
77628-798EA 1484.23 USD
77628-798 77638-324 77650-346
6-Bromo-5-fluoro-3-iodo-1-methyl-1H-indazole ≥95%
6-Bromo-5-fluoro-3-iodo-1-methyl-1H-indazole

Precaución: For Research Use Only

Formula: C₈H₅BrFIN₂
MW: 354.946 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384874
CAS Number: 1286734-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR