Cyclohexyl(2-fluoro-5-iodophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

357743-5G 357743-25G 357743-1G 357743-500MG
77628-794EA 1705.68 USD
77628-794 77635-698 77638-322 77650-342
Cyclohexyl(2-fluoro-5-iodophenyl)methanone ≥95%
Cyclohexyl(2-fluoro-5-iodophenyl)methanone

Precaución: For Research Use Only

Formula: C₁₃H₁₄FIO
Storage Temperature: Ambient
MDL Number: MFCD35190998
CAS Number: 3002493-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR