6-(Tert-butylthio)-2-fluoro-3-iodopyridine ≥95%

Supplier: AOBChem USA

Warning

356888-5G 356888-25G 356888-1G 356888-500MG
77628-790EA 1484.23 USD
77628-790 77635-694 77642-332 77650-340
6-(Tert-butylthio)-2-fluoro-3-iodopyridine ≥95%
6-(Tert-butylthio)-2-fluoro-3-iodopyridine

Precaución: For Research Use Only

Formula: 311.1582 g/mol
MW: C₉H₁₁FINS
Storage Temperature: Ambient
MDL Number: MFCD35190582
CAS Number: 3002440-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR