6-(Tert-butylthio)-2-fluoronicotinic acid ≥95%

Supplier: AOBChem USA

Warning

356881-5G 356881-25G 356881-1G 356881-500MG
77628-786EA 1484.23 USD
77628-786 77635-692 77642-330 77646-342
6-(Tert-butylthio)-2-fluoronicotinic acid ≥95%
6-(Tert-butylthio)-2-fluoronicotinic acid

Precaución: For Research Use Only

Formula: C₁₀H₁₂FNO₂S
Storage Temperature: Ambient
MDL Number: MFCD35190575
CAS Number: 3002431-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR