3,5-Dichloro-4-iodoaniline ≥97%

Supplier: AOBChem USA

Warning

356821-5G 356821-25G 356821-1G 356821-500MG
77632-916EA 1356 USD
77632-916 77655-354 77642-328 77650-338
3,5-Dichloro-4-iodoaniline ≥97%
3,5-Dichloro-4-iodoaniline

Precaución: For Research Use Only

Formula: C₆H₄Cl₂IN
MW: 287.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20926090
CAS Number: 155403-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR