5-Bromo-6-ethylpyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

356737-5G 356737-1G 356737-500MG
77628-782EA 1484.23 USD
77628-782 77638-316 77650-336
5-Bromo-6-ethylpyridin-2-amine ≥97%
5-Bromo-6-ethylpyridin-2-amine

Precaución: For Research Use Only

Formula: C₇H₉BrN₂
Storage Temperature: Ambient
CAS Number: 848360-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR