Cyclopropyl(3-ethoxy-2-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

356463-5G 356463-25G 356463-1G 356463-500MG
77628-780EA 1484.23 USD
77628-780 77635-690 77642-326 77650-334
Cyclopropyl(3-ethoxy-2-methylphenyl)methanone ≥95%
Cyclopropyl(3-ethoxy-2-methylphenyl)methanone

Precaución: For Research Use Only

Formula: 204.265 g/mol
MW: C₁₃H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD35190359
CAS Number: 3002542-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR