6-Bromo-3-chloro-α-ethenyl-2-fluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

355996-1G 355996-5G 355996-500MG
77638-308EA 537 USD
77638-308 77658-912 77646-336
6-Bromo-3-chloro-α-ethenyl-2-fluorobenzenemethanol ≥95%
6-Bromo-3-chloro-α-ethenyl-2-fluorobenzenemethanol

Precaución: For Research Use Only

Formula: C₉H₇BrClFO
Storage Temperature: Ambient
MDL Number: MFCD35189951
CAS Number: 2228189-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR