α-Ethenyl-2,4,6-trifluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

355966-5G 355966-1G 355966-500MG
77655-348EA 1921 USD
77655-348 77642-322 77646-334
α-Ethenyl-2,4,6-trifluorobenzenemethanol ≥95%
α-Ethenyl-2,4,6-trifluorobenzenemethanol

Precaución: For Research Use Only

Formula: C₉H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD35189930
CAS Number: 1599101-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR