3-Chloro-α-ethenyl-2,6-difluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

355938-5G 355938-1G 355938-500MG
77628-772EA 1484.23 USD
77628-772 77638-304 77650-326
3-Chloro-α-ethenyl-2,6-difluorobenzenemethanol ≥97%
3-Chloro-α-ethenyl-2,6-difluorobenzenemethanol

Precaución: For Research Use Only

Formula: C₉H₇ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD35189923
CAS Number: 1422509-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR