2-(3,5-Dimethylphenoxy)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

355541-5G 355541-1G 355541-25G 355541-500MG
77628-770EA 1718.04 USD
77628-770 77638-302 77655-346 77646-332
2-(3,5-Dimethylphenoxy)benzonitrile ≥97%
2-(3,5-Dimethylphenoxy)benzonitrile

Precaución: For Research Use Only

Formula: C₁₅H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD11125400
CAS Number: 1040310-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR