2-(4-Fluorophenoxy)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

355537-5G 355537-25G 355537-1G 355537-500MG
77634-914EA 602 USD
77634-914 77658-910 77642-320 77646-330
2-(4-Fluorophenoxy)benzonitrile ≥97%
2-(4-Fluorophenoxy)benzonitrile

Precaución: For Research Use Only

Formula: C₁₃H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD11155956
CAS Number: 1020922-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR